Thermo Scientific Nalgene 3120-1010 Centrifuge bottle; 1000 mL; PPCO; for IEC rotor, 4/pk
MFLX06107-75
- Catalog No:
- MFLX06107-75
- MFLX06106-40
- MFLX06105-60
- Tube capacity:
- 100 ml
- 100 ml
- 100 ml
- Autoclavable:
- Yes
- Yes
- Yes
- Tube bottom shape:
- Round
- Round
- Round
- Max. RCF:
- 7100 ×g
- 7100 ×g
- 7100 ×g
- Tube ext.Ø:
- 97,9 mm
- 97,6 mm
- 97,5 mm
Get better separation of large samples.
- Autoclavable
Choose from PPCO or PC. Bottles are injection blown—no “pinchmarks” that could contribute to failure. To use bottles at maximum speeds without leakage or to spin hazardous materials, order sealing caps under accessories. Models 06106-40 and 06105-60 fit the following rotors: Beckman JS-5.2, JS-4.2, JS-4.0, and JS-3.0 (when used with Beckman 344040 adapter). All models fit Sorvall HG-4L, H-4000, H-6000A (no adapter needed) and IEC 266, 949, and 966 (when used with appropriate IEC adapter).
Warranty: 1 year
- Cap included:Yes
- Cap material:PP
- Colour:Natural
- Description:Centrifuge bottle; 1000 mL; PPCO; for IEC rotor, 4/pk
- Length:179 mm
- Material:PP
- Environmentally Preferable:
- Tube bottom shape:Round
- Tube capacity:100 ml
- Max. RCF:7100 ×g
- Tube ext.Ø:97,9 mm
- Autoclavable:Yes
- Pk:4
- Closure size no.:63
Frequently Bought Together