Labnet Hermle C1205 Tube Adapters, 0.5/0.6 mL; 6/Pk
Catalog # MFLX17707-13
Supplier: Avantor Fluid Handling
About this item
Choose the centrifuge tube adapter to suit your application needs!
- Expand and change the capacity of your centrifuge with these adapters
Customize your Labnet centrifuge with these tube adapters.
- Description:Tube Adapters, 0,5/0,6 mL; 6/Pk
- Environmentally Preferable:
- Pk:6
- No. of tubes accepted:1
- Tube sizes accepted:0,5/0,6 ml
Frequently Bought Together