Thermo Scientific Nunc Conical-Bottom Centrifuge Tubes, PC, 15 mL; 10/PK
MFLX06326-10
Molded-in graduation simplify measurements.
- Designed for use up to 6000 × g in refrigerated or non-refrigerated centrifuges
- Autoclavable
The conical-bottle centrifuge tubes feature molded-in graduations allowing you to easily read measurements. They are designed for low-speed centrifugation up to 6000 ×g in refrigerated or non-refrigerated centrifuges. Please note, tubes must be filled to at least 80% capacity for proper function. The conical bottoms concentrate pellet in a small area for easy isolation and retrieval. Available in either polypropylene copolymer (PPCO) or polycarbonate (PC).
The translucent PPCO tubes are compatible with a wide range of lab reagents and offer excellent chemical resistance. The PC tubes feature outstanding mechanical strength and clarity.
- Cap included:No
- Tube ext.Ø:16,9 mm
- Colour:Clear
- Description:Conical-bottom tubes; 15 mL; PC; 10/pack
- Length:120,2 mm
- Material:Polycarbonate
- Environmentally Preferable:
- Warranty:1 year
- Autoclavable:Yes
- Graduations:+
- Pk:10
- Tube bottom shape:Conical
- Tube capacity:15 ml
Frequently Bought Together