Thermo Scientific Nalgene 3141-0250 Centrifuge Bottle, Sealing, PPCO, 250 mL; 4/Pk
Catalog # MFLX06106-14
Supplier: Avantor Fluid Handling
About this item
Get better separation of large samples.
- Measure more precisely with molded-in graduations
- Leakproof
- Include leakproof, PP cap
The bottles are available in HDPE, FEP, PPCO or PC. Feature molded-in graduations and PP sealing cap.
Use model 06107-20 at low speeds, filled completely, and in refrigerated centrifuge to maintain leak-free spinning. Bottles 06107-20 and 06106-10 require sealing cap assembly 06327-57 for spinning at high speeds.
06107-00 require the noted adapter to fit most rotors; order adapters separately. 06105-20, 06107-10, 06107-20, 06106-10, 06106-12, 06106-14 require the noted adapter to fit the Sorvall/Hereaus GSA rotor; order adapters separately. In order to reach maximum speed ratings without leaking or to spin hazardous materials, order 38-mm sealing cap assembly 06327-57, or 63-mm sealing cap 06327-69.
- Cap included:Yes
- Cap material:PP
- Capacity:250 ml
- Colour:Natural
- Description:Centrifuge Bottle, Sealing, PPCO; 4/Pk
- Length:133,4 mm
- Material:PPCO
- Environmentally Preferable:
- Tube bottom shape:Flat
- Tube capacity:250 ml
- Max. RCF:27500 ×g
- Tube ext.Ø:60,7 mm
- Autoclavable:Yes
- Pk:4
- Closure size no.:58
Frequently Bought Together