Thermo Scientific Nalgene 3120-0250 Centrifuge bottle; 250 mL; PPCO; 4/pack
MFLX06105-20
Get better separation of large samples.
- Measure more precisely with molded-in graduations
- Leakproof
- Include leakproof, PP cap
The bottles are available in HDPE, FEP, PPCO or PC. Feature molded-in graduations and PP sealing cap.
Use model 06107-20 at low speeds, filled completely, and in refrigerated centrifuge to maintain leak-free spinning. Bottles 06107-20 and 06106-10 require sealing cap assembly 06327-57 for spinning at high speeds.
06107-00 require the noted adapter to fit most rotors; order adapters separately. 06105-20, 06107-10, 06107-20, 06106-10, 06106-12, 06106-14 require the noted adapter to fit the Sorvall/Hereaus GSA rotor; order adapters separately. In order to reach maximum speed ratings without leaking or to spin hazardous materials, order 38-mm sealing cap assembly 06327-57, or 63-mm sealing cap 06327-69.
- Cap included:Yes
- Cap material:PP
- Capacity:250 ml
- Colour:Natural
- Description:Centrifuge bottle; PPCO; 4/pack
- Length:127,7 mm
- Material:PP
- Environmentally Preferable:
- Tube bottom shape:Round
- Tube capacity:250 ml
- Max. RCF:13200 ×g
- Tube ext.Ø:61,8 mm
- Autoclavable:Yes
- Pk:4
- Closure size no.:38
Frequently Bought Together