Tubing and Connectors, 3/16" ID
Catalog # MFLX98767-99
Supplier: Avantor Fluid Handling
About this item
Choose the size or assembly compatible with your pump drive.
- Choose the flexible tubing or tubing set with connectors to replace spent, old, or broken tubing
Replacement tube sets are designed specifically for use in the Variable-Speed Peristaltic Pumps.
- Colour:Translucent
- Description:Tubing and Connectors, 3/16" ID
- Environmentally Preferable:
- Warranty:1 year
- Clarity:Translucent
- Tubing material:Silicone
- Pk:1
Frequently Bought Together