In-line Filter 35um PEEK for Dosing Pumps 78167-00 to -07
Catalog # MFLX78167-51
Supplier: Avantor Fluid Handling
1 year warranty
About this item
Replacement parts keep your dosing pump running smoothly.
- Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
- Description:In-line Filter 35um PEEK for Dosing Pumps 78167-00 to -07
- Environmentally Preferable:
- Wetted materials:PEEK
- Warranty:1 year
- Pk:1
Frequently Bought Together