Neptune Feed Verification Diaphragm Pump Flow Checkers
Flow checker; for models 73803-00, -30, -40; 73804-00, -30, -40
Catalog # MFLX73805-00
Supplier: Avantor Fluid Handling
Neptune Feed Verification Diaphragm Pump Flow Checkers
Catalog # MFLX73805-00
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Easily meansure flow of your Feed Verification Diaphragm Pump.
- Optimize performance of your Neptune Metering Pumps
These flow checkers can be mounted directly onto the discharge end of the pumps. The checker is a simple oval gear meter that measures the pump output and then transmits one pulse for every one ml of flow. It requires 4.5 to 25 VDC at a 20 mA maximum. Pump sold separately.
Specifications
- Description:Flow checker; for models 73803-00, -30, -40; 73804-00, -30, -40
- Cat. No.:MFLX73805-00
- Warranty:1 year