Water Pump, 6 strokes/gallon, 3 foot inlet tube
MFLX70607-14
- Flow rate:6 strokes/gal.
- Description:Water Pump, 6 strokes/gallon, 3 foot inlet tube
- Environmentally Preferable:
- Wetted materials:PVC body, hose and shaft
- Tube length, inlet:36"
- Warranty:1 Month
- Pk:1
Ideal for removing water in municipal and irrigation markets in addition to facilities maintenance.
- Recommended for aqueous solutions
- Withstand temperatures up to 120F (49C)
- Self-priming, will not siphon
This hand-operated drum pump is ideal for pumping water and aqueous solutions. Constructed with a PVC body, hose and shaft, this pump offers 6 strokes/gallon.
- Flow rate:6 strokes/gal.
- Description:Water Pump, 6 strokes/gallon, 3 foot inlet tube
- Environmentally Preferable:
- Wetted materials:PVC body, hose and shaft
- Tube length, inlet:36"
- Warranty:1 Month
- Pk:1
Frequently Bought Together