Order Entry
India
ContactUsLinkComponent
Bushing, PTFE, 3/8" NPT(M) X 1/4" NPT(F)
Bushing, PTFE, 3/8" NPT(M) X 1/4" NPT(F)
Catalog # MFLX34030-45
undefined
Bushing, PTFE, 3/8" NPT(M) X 1/4" NPT(F)
Catalog # MFLX34030-45
About this item
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings offer more permanent installation. Choose the size and material that meets your application needs.
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 2):
    ¼"
  • Thread size NPT(M) (port 1):
    ⅜"
  • Description:
    Bushing, PTFE, ⅜" NPT(M) × ¼" NPT(F)
  • Seq:
    42
  • Environmentally Preferable:
  • Model:
    34030-45
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Pk:
    1
  • Fitting material:
    PTFE
Frequently Bought Together