McMillan Flow Power supply/input/output cable (0 to 5 VDC), 115 VAC
Catalog # MFLX32708-50
Supplier: Avantor Fluid Handling
About this item
Make the most of your McMillian Flow flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
Replacement cables, power supplies, and other accessories are suitable for use with your McMillian Flow flowmeters.
- Description:Power supply/input/output cable (0 to 5 VDC), 115 VAC
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
Frequently Bought Together