Order Entry
India
ContactUsLinkComponent
McMillan Flow 100-17T Ryton and Brass Sensors Cable assembly, 30" (0.9 m) L
McMillan Flow 100-17T Ryton and Brass Sensors Cable assembly, 30" (0.9 m) L
Catalog # MFLX32704-52
CE Compliant
undefined
McMillan Flow 100-17T Ryton and Brass Sensors Cable assembly, 30" (0.9 m) L
Catalog # MFLX32704-52
CE Compliant

About this item

Make the most of your McMillian Flow flowmeters with these replacement parts and other accessories.

  • Optimize performance of your flowmeter with these accessories

Replacement cables, power supplies, and other accessories are suitable for use with your McMillian Flow flowmeters.

  • Description:
    Ryton and Brass Sensors Cable assembly, 30" (0,9 m) L
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together