Parker Hannifin Fitting, PFA, Tee, Compression to Thread, 1/2" OD x 1/2" OD x 1/4" NPT(M)
Catalog # MFLX31321-80
Supplier: Avantor Fluid Handling
About this item
Securely connect your rigid tubing.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
These branch tees feature a male NPT thread on the bottom branch for quick connection to a threaded port.
- Connection Type:Compression to Thread
- Flow pattern:T
- Description:Fitting, PFA, Tee, Compression Branch to Threaded Adapter, ½" OD x ½" OD x ¼" NPT(M)
- Environmentally Preferable:
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Warranty:18 months
- Pk:1
- Fitting material:PFA
Frequently Bought Together