Replacement PFA Compression Ferrule Nut and Assemblies
Ferrule plug, PFA, ¼", Compression Plug, PFA, _
MFLX31320-46
CE certificate
Replacement PFA Compression Ferrule Nut and Assemblies
MFLX31320-46
CE certificate
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
- Connection Type:Compression Plug
- Flow pattern:_
- Description:Ferrule plug, PFA, ¼"
- Fitting material:PFA
- Cat. No.:MFLX31320-46
- Warranty:18 months
- Sterilisation:Autoclave or ethylene oxide (ETO)