Parker Hannifin Flaring tool for tubing 1/4" outer diameter
Catalog # MFLX30490-03
Supplier: Avantor Fluid Handling
About this item
Chemically-inert PFA handles most aggressive chemicals.
- Optimize performance of your PFA Flare Fittings
Use these tools for a better fit. Heat the PFA tubing with a heat gun, and insert the tool to create the flare. Tools are available for any size of tubing from 1/4-inch to 1-inch outer diameter.
- Connection Type:Flare Union
- Flow pattern:Straight
- Description:Flaring tool for tubing ¼" outer diameter
- Environmentally Preferable:
- Warranty:3 Months
- Pk:1
- Fitting material:PFA
Frequently Bought Together