Order Entry
India
ContactUsLinkComponent
American Machine & Tool Motor Foot Bracket for NEMA Type 56 C-Face Motors
American Machine & Tool Motor Foot Bracket for NEMA Type 56 C-Face Motors
Catalog # MFLX07127-80
undefined
American Machine & Tool Motor Foot Bracket for NEMA Type 56 C-Face Motors
Catalog # MFLX07127-80

About this item

Create your own motor mounting with the hardware below.

  • Easily mount your American Machine & Tool Motor NEMA Type 56 C-face motors
  • Provides base for NEMA type 56 C-face motors

Motor Foot Bracket provides a base for NEMA Type 56 C-face motors.

  • Description:
    Motor foot bracket, provides base for NEMA type 56 C-face motors
  • Length:
    15,24 cm
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
  • Width:
    7,62 cm
  • Motor frame size:
    56C
Frequently Bought Together