Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
Catalog # MFLX07079-15
Supplier: Avantor Fluid Handling
About this item
Compatible with Siphon and Piston Hand-Operated Pumps.
- Choose from a wide range of replacement parts and accessories
Model 07079-10 steel screw in adapter features a 1-3/4" NPT(F) connection and fits 5 gallon pails.
Model 07079-15 flex spout adapter is constructed of durable polyethylene and fits 5 gallon containers with 5-5/8" (6.7 cm) openings.
Model 07079-17 bung converter kit converts a Mauser course bung thread or standard course buttress to a 2" NPT connection.
- Description:Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
Frequently Bought Together