Parker Hannifin Compression Fitting Ferrule, 316 Stainless Steel, 1/8" OD; 1/EA
MFLX03302-26
Get a secure seal at higher pressures.
- Constructed of durable stainless steel
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Ferrules are required to create a leak-tight seal.
- Connection Type:Compression Union
- Flow pattern:Straight
- Description:Compression Fitting Ferrule, 316 Stainless Steel, ⅛" OD; 1/EA
- Environmentally Preferable:
- Sterilisation:Ethylene oxide (EtO), autoclave, or gamma irradiation
- Warranty:1 year
- Pk:1
- Fitting material:Stainless Steel
Frequently Bought Together