Masterflex® Fitting Clamp, PVDF, Sanitary Tri-Clamp®, 1/2"; 1/EA
Catalog # MFLX31201-88
Supplier: Avantor Fluid Handling
About this item
Maintain sanitary conditions in your processes.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These sanitary clamps are compatible with Tri-Clamp® fittings. The PVDF material has excellent chemical resistance with low leachable characteristics.
- Connection type:Sanitary
- Description:Push/pull clamp; PVDF for sanitary fittings
- Material:PVDF
- Environmentally Preferable:
- Model:31201-88
- Sanitary clamp size (port 1):1/2"
- Pk:1
- Clamp type:Sanitary
Frequently Bought Together