PendoTech PDKT-650-298MG Single-Use Cable with RJ12 to Midgee Monitor; 6' (1.8 m)
MFLX19406-86
CE Compliant
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
- Length:182,88 cm (72")
- Description:Cable with RJ12 to Midgee Monitor, 6 ft (1,8 m)
- Environmentally Preferable:
- Warranty:2 years
- Pk:1
Frequently Bought Together