Order Entry
India
ContactUsLinkComponent
Tubing Rack, 10-Channel for 3/16" OD Tubing, Non-Adhesive Back, 10/Pk
Tubing Rack, 10-Channel for 3/16" OD Tubing, Non-Adhesive Back, 10/Pk
Catalog # MFLX06432-02
undefined
Tubing Rack, 10-Channel for 3/16" OD Tubing, Non-Adhesive Back, 10/Pk
Catalog # MFLX06432-02

About this item

An easy solution to secure and route multiple tubing lines.

  • Keep your tubing lines organised and untangled

These tube racks offer a simple and economical method for organising tubing lines. The ³/₁₆" and ¹/₄" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The ³/₁₆" and ¹/₄" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; ³/₁₆" and ¹/₄" sizes are available in other colours.

  • Description:
    Tubing Rack, 10-Channel for 3/16" OD Tubing, Non-Adhesive Back, 10/Pk
  • Seq:
    03
  • Environmentally Preferable:
  • No. of channels:
    10
  • Warranty:
    1 year
  • Pk:
    10
  • For tubing ext.Ø:
    4,76 mm (0,1875")
Frequently Bought Together