Order Entry
India
ContactUsLinkComponent
Gentamicin Sulfate Reagent Solution is recommended for use in cell culture applications at 5 ml/L and used as a selection agent (gen tamicin-resistance gene) in molecular biology applications 1 * 20 mL
Catalog # VWRA06210003
Supplier:  VWR Collection
Gentamicin Sulfate Reagent Solution is recommended for use in cell culture applications at 5 ml/L and used as a selection agent (gen tamicin-resistance gene) in molecular biology applications 1 * 20 mL
Catalog # VWRA06210003
Supplier:  VWR Collection
Frequently Bought Together