Order Entry
India
ContactUsLinkComponent
Kanamycin Monosulfate used in biotechnology applications to inhibit protein synthesis. It is also used as a selection agent for cell s transformed with kanamycin B (neoR, kanR) resistance gene 1 * 100 g
Catalog # VWRA06201105
Supplier:  VWR Collection
Kanamycin Monosulfate used in biotechnology applications to inhibit protein synthesis. It is also used as a selection agent for cell s transformed with kanamycin B (neoR, kanR) resistance gene 1 * 100 g
Catalog # VWRA06201105
Supplier:  VWR Collection
Frequently Bought Together