Order Entry
India
ContactUsLinkComponent
D-Mannitol used with boric acid in the manufacturing of dry electrolytic condensers for radio applications. In pharmacy as excipient and diluent for solids and liquids. In analytical chemistry for boron determinations. In the manufuring of mannitol s 1 * 500 g
Catalog # VWRA06190128
Supplier:  VWR Collection
D-Mannitol used with boric acid in the manufacturing of dry electrolytic condensers for radio applications. In pharmacy as excipient and diluent for solids and liquids. In analytical chemistry for boron determinations. In the manufuring of mannitol s 1 * 500 g
Catalog # VWRA06190128
Supplier:  VWR Collection
Frequently Bought Together