Order Entry
India
ContactUsLinkComponent
Sodium pyruvate is utilized as a component in culture broth and media. The use of sodium pyruvate in Wallen fermentation medium to e nhance the conversion of oleic acid to 10-ketostearic acid by Bacillus sphaericus has been described. A protocol that 1 * 250 g
Catalog # VWRA06190125
Supplier:  VWR Collection
Sodium pyruvate is utilized as a component in culture broth and media. The use of sodium pyruvate in Wallen fermentation medium to e nhance the conversion of oleic acid to 10-ketostearic acid by Bacillus sphaericus has been described. A protocol that 1 * 250 g
Catalog # VWRA06190125
Supplier:  VWR Collection
Frequently Bought Together