Order Entry
India
ContactUsLinkComponent
Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions, particularly in pharm aceutical applications. It has been used in pre-extraction of membranes to remove peripheral proteins. It has been us 1 * 1.000 mL
Catalog # VWRA06190123
Supplier:  VWR Collection
Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions, particularly in pharm aceutical applications. It has been used in pre-extraction of membranes to remove peripheral proteins. It has been us 1 * 1.000 mL
Catalog # VWRA06190123
Supplier:  VWR Collection
Frequently Bought Together