Order Entry
India
ContactUsLinkComponent
Silver nitrate occurs naturally as the mineral ore acanthite. Silver nitrate is a source of silver ions in biological studies, such as in toxicological investigations on yeast and marine animals. It has been used to induce the protein conformational 1 * 100 g
Catalog # VWRA06190121
Supplier:  VWR Collection
Silver nitrate occurs naturally as the mineral ore acanthite. Silver nitrate is a source of silver ions in biological studies, such as in toxicological investigations on yeast and marine animals. It has been used to induce the protein conformational 1 * 100 g
Catalog # VWRA06190121
Supplier:  VWR Collection
Frequently Bought Together