Order Entry
India
ContactUsLinkComponent
Tricine can be used in cryopreservation medium for the preservation of tissues and organs. Cryopreservation depends on the physical and chemical characteristics of the preservation medium used. Tricine has been found to be an efficient scavenger of h 1 * 500 g
Catalog # VWRA06190119
Supplier:  VWR Collection
Tricine can be used in cryopreservation medium for the preservation of tissues and organs. Cryopreservation depends on the physical and chemical characteristics of the preservation medium used. Tricine has been found to be an efficient scavenger of h 1 * 500 g
Catalog # VWRA06190119
Supplier:  VWR Collection
Frequently Bought Together