Order Entry
India
ContactUsLinkComponent
Buffers can be made by adding a solution of base to PIPES free acid, titrating to the appropriate pH, or by mixing equimolar solutio ns of the sesquisodium salt and the disodium salt or dipotassium salt, titrating to the appropriate pH. 1 * 100 g
  VWRA06190117
Buffers can be made by adding a solution of base to PIPES free acid, titrating to the appropriate pH, or by mixing equimolar solutio ns of the sesquisodium salt and the disodium salt or dipotassium salt, titrating to the appropriate pH. 1 * 100 g
  VWRA06190117
Frequently Bought Together