Order Entry
India
ContactUsLinkComponent
An irreversible inhibitor of serine proteases - inhibits trypsin, chymotrypsin and protease. Causes sulfonylation of the active-site serine residue. A less toxic substitute for the extremely toxic diisopropylfluorophosphate. Inhibits mammalian acety 1 * 25 g
Catalog # VWRA06190116
Supplier:  VWR Collection
An irreversible inhibitor of serine proteases - inhibits trypsin, chymotrypsin and protease. Causes sulfonylation of the active-site serine residue. A less toxic substitute for the extremely toxic diisopropylfluorophosphate. Inhibits mammalian acety 1 * 25 g
Catalog # VWRA06190116
Supplier:  VWR Collection
Frequently Bought Together