Order Entry
India
ContactUsLinkComponent
5-Bromo-4-Chloro-3-Indolyl--D-Galactopyranoside is used as indigogenic substrate for -galactosidase, for detection of -galactosid ase-positive clones, and the identification of lac and bacterial colonies or phage plaques. It is the substrate of ch 1 * 100 mg
Catalog # VWRA06104116
Supplier:  VWR Collection
5-Bromo-4-Chloro-3-Indolyl--D-Galactopyranoside is used as indigogenic substrate for -galactosidase, for detection of -galactosid ase-positive clones, and the identification of lac and bacterial colonies or phage plaques. It is the substrate of ch 1 * 100 mg
Catalog # VWRA06104116
Supplier:  VWR Collection
Frequently Bought Together