Order Entry
India
ContactUsLinkComponent
Ethidium bromide is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis. The fluorescence of EtBr incr eases 21-fold upon binding to double-stranded RNA and 25-fold on binding double-stranded DNA so that destaining the b 1 * 10 g
Catalog # VWRA06104118
Supplier:  VWR Collection
Ethidium bromide is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis. The fluorescence of EtBr incr eases 21-fold upon binding to double-stranded RNA and 25-fold on binding double-stranded DNA so that destaining the b 1 * 10 g
Catalog # VWRA06104118
Supplier:  VWR Collection
Frequently Bought Together