Order Entry
India
ContactUsLinkComponent
Zwitterionic amino acid which acts as one of the Good biological buffer,More suitable as a biological buffer than MES because its pH range is closer to pH 7.4 1 * 500 g
Catalog # VWRA06104107
Supplier:  VWR Collection
Zwitterionic amino acid which acts as one of the Good biological buffer,More suitable as a biological buffer than MES because its pH range is closer to pH 7.4 1 * 500 g
Catalog # VWRA06104107
Supplier:  VWR Collection
Frequently Bought Together