Order Entry
India
ContactUsLinkComponent
Glycerol is used in the concentration and storage of enzymes. Also prevents back-diffusion and protein samples into the buffer. Glyc erol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) in 1 * 1.000 mL
Catalog # VWRA06104105
Supplier:  VWR Collection
Glycerol is used in the concentration and storage of enzymes. Also prevents back-diffusion and protein samples into the buffer. Glyc erol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) in 1 * 1.000 mL
Catalog # VWRA06104105
Supplier:  VWR Collection
Frequently Bought Together