Order Entry
India
ContactUsLinkComponent
Boric Acid, Molecular Biology Grade, in conjunction with Tris, is commonly used in buffers for the preparation of agarose or acrylam ide gels and their associated running buffers 1 * 500 g
Catalog # VWRA06104102
Supplier:  VWR Collection
Boric Acid, Molecular Biology Grade, in conjunction with Tris, is commonly used in buffers for the preparation of agarose or acrylam ide gels and their associated running buffers 1 * 500 g
Catalog # VWRA06104102
Supplier:  VWR Collection
Frequently Bought Together