Order Entry
India
ContactUsLinkComponent
LABORATORY CENTRIFUGE NEOFUGE 13R, REFRIGERATION FUNCTION,13800 MAX RPM, 17459xg MAX RCF, WITH 2201 ANGLE ROTOR AS STANDARD Laboratory centrifuge neofuge 13R, refrigeration function,13800 max RPM, 17459XG max RCF, with 2201 angle rotor as standard 1 * 1 items
Catalog # HEALNEOFUGE13R
Supplier:  HEAL FORCE
LABORATORY CENTRIFUGE NEOFUGE 13R, REFRIGERATION FUNCTION,13800 MAX RPM, 17459xg MAX RCF, WITH 2201 ANGLE ROTOR AS STANDARD Laboratory centrifuge neofuge 13R, refrigeration function,13800 max RPM, 17459XG max RCF, with 2201 angle rotor as standard 1 * 1 items
Catalog # HEALNEOFUGE13R
Supplier:  HEAL FORCE
Frequently Bought Together