Order Entry
India
ContactUsLinkComponent
LABORATORY CENTRIFUGE NEOFUGE 15, 16000 MAX RPM, 24900xg MAX RCF, WITH 1501 ANGLE ROTOR AS STANDARD Laboratory centrifuge neofuge 15, 16000 max RPM, 24900XG max RCF, with 1501 angle rotor as standard 1 * 1 items
Catalog # HEALNEOFUGE15
Supplier:  HEAL FORCE
LABORATORY CENTRIFUGE NEOFUGE 15, 16000 MAX RPM, 24900xg MAX RCF, WITH 1501 ANGLE ROTOR AS STANDARD Laboratory centrifuge neofuge 15, 16000 max RPM, 24900XG max RCF, with 1501 angle rotor as standard 1 * 1 items
Catalog # HEALNEOFUGE15
Supplier:  HEAL FORCE
Frequently Bought Together