Order Entry
India
ContactUsLinkComponent
LABORATORY CENTRIFUGE NEOFUGE 15R, REFRIGERATION FUNCTION,16000 MAX RPM, 24900xg MAX RCF, WITH 1501 ANGLE ROTOR AS STANDARD Laboratory centrifuge neofuge 15R, refrigeration function,16000 max RPM, 24900XG max RCF, with 1501 angle rotor as standard 1 * 1 items
Catalog # HEALNEOFUGE15R
Supplier:  HEAL FORCE
LABORATORY CENTRIFUGE NEOFUGE 15R, REFRIGERATION FUNCTION,16000 MAX RPM, 24900xg MAX RCF, WITH 1501 ANGLE ROTOR AS STANDARD Laboratory centrifuge neofuge 15R, refrigeration function,16000 max RPM, 24900XG max RCF, with 1501 angle rotor as standard 1 * 1 items
Catalog # HEALNEOFUGE15R
Supplier:  HEAL FORCE
Frequently Bought Together