Order Entry
India
ContactUsLinkComponent
CO2 INCUBATOR MODEL HF90, AIR JACKETED, DIRECT HEAT, 151L INTERNAL CAPACITY, 3 DESIGNED INNER DOORS CO2 incubator model HF90, air jacketed, direct heat, 151L internal capacity, 3 designed inner doors 1 * 1 items
Catalog # HEALHF90-3ID
Supplier:  HEAL FORCE
CO2 INCUBATOR MODEL HF90, AIR JACKETED, DIRECT HEAT, 151L INTERNAL CAPACITY, 3 DESIGNED INNER DOORS CO2 incubator model HF90, air jacketed, direct heat, 151L internal capacity, 3 designed inner doors 1 * 1 items
Catalog # HEALHF90-3ID
Supplier:  HEAL FORCE
Frequently Bought Together