Order Entry
India
ContactUsLinkComponent
TRI-GAS INCUBATOR MODEL HF100, AIR JACKETED, DIRECT HEAT, CO2/O2 CONTROL, 151L INTERNAL CAPABILITY Tri-gas incubator model HF100, air jacketed, direct heat, CO2/O2 control, 151L internal capability 1 * 1 items
Catalog # HEALHF100
Supplier:  HEAL FORCE
TRI-GAS INCUBATOR MODEL HF100, AIR JACKETED, DIRECT HEAT, CO2/O2 CONTROL, 151L INTERNAL CAPABILITY Tri-gas incubator model HF100, air jacketed, direct heat, CO2/O2 control, 151L internal capability 1 * 1 items
Catalog # HEALHF100
Supplier:  HEAL FORCE
Frequently Bought Together