J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
4, ±0,04, Phthalate, Red, 25 °C, Plastic bottle, 1 L
Supplier: Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Catalog # 5657-02
Supplier: Avantor
CAS Number:
Jump to:
Product Details & Documents
The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).
- Colour coded
- Potassium hydrogen phthalate based pH 4 buffer (red)
- Phosphate based pH 7 buffer (yellow)
- Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and colour-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Each lot compared to NIST Standard Reference Buffer.
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phthalate
- Pk:1 L
- Reference temperature:25 °C
- Colour:Red
- pH accuracy:±0,04
- pH value:4



