Order Entry
India
ContactUsLinkComponent
OMNIA TAP UV6 PURE WATER GENERATION IN TANK AT 15C 6L/H SYSTEM HAS A BUILT IN TANK OF 10 LTR, CONDUCTIVITY 0.055 ?S/CM DISPENSING PERFORMANCE UP TO 2 L/MIN. TOC VALUE 1 - 5 PPB PARTICLE 0.2 M < 1/ML DISPENSE @ 2LTR/MIN. 1 * 1 items
Catalog # IBSA18200052
Supplier:  IBS STOCK ITEMS
OMNIA TAP UV6 PURE WATER GENERATION IN TANK AT 15C 6L/H SYSTEM HAS A BUILT IN TANK OF 10 LTR, CONDUCTIVITY 0.055 ?S/CM DISPENSING PERFORMANCE UP TO 2 L/MIN. TOC VALUE 1 - 5 PPB PARTICLE 0.2 M < 1/ML DISPENSE @ 2LTR/MIN. 1 * 1 items
Catalog # IBSA18200052
Supplier:  IBS STOCK ITEMS
Frequently Bought Together