Order Entry
India
ContactUsLinkComponent
Maxi Sub XL System: Extra Large Model (Custom built) Gel Size 30cmx25cm (for 33x3=99 samples) with inbuilt comb stand Multi Channel Pipette Compatible 1 * 1 items
Catalog # GNEI6614GB
Supplier:  GENEI LABORATORIE
Maxi Sub XL System: Extra Large Model (Custom built) Gel Size 30cmx25cm (for 33x3=99 samples) with inbuilt comb stand Multi Channel Pipette Compatible 1 * 1 items
Catalog # GNEI6614GB
Supplier:  GENEI LABORATORIE
Frequently Bought Together