Rs.0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
India
ContactUsLinkComponent
Taq DNA Polymerase (5 U/µl) (Includes Enzyme: 1 vial 10X containing TritonX-100: 4 vials 25 mM MgCl2: 4 vials), 1000Units 1 * 1 KU
GNEI0603300051730
:
GENEI LABORATORIE
Taq DNA Polymerase (5 U/µl) (Includes Enzyme: 1 vial 10X containing TritonX-100: 4 vials 25 mM MgCl2: 4 vials), 1000Units 1 * 1 KU
GNEI0603300051730
:
GENEI LABORATORIE
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals