Order Entry
India
ContactUsLinkComponent
Refrigerated Circulator / Chiller with Temp. Range : -20 0C to RT, Heater : 2000 watt, Flow Rate (Max.) : 12 lit / min, Pump Pressure : 300mbar, Temperature Accuracy : +/-0.1 0C, Refrigerant : CFC free, Cooling Capacity @20 0C: 300 watt,Bath Material 1 * 1 items
  462-0602
Refrigerated Circulator / Chiller with Temp. Range : -20 0C to RT, Heater : 2000 watt, Flow Rate (Max.) : 12 lit / min, Pump Pressure : 300mbar, Temperature Accuracy : +/-0.1 0C, Refrigerant : CFC free, Cooling Capacity @20 0C: 300 watt,Bath Material 1 * 1 items
  462-0602
Frequently Bought Together