J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Product Details & Documents
The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).
- Colour coded
- Potassium hydrogen phthalate based pH 4 buffer (red)
- Phosphate based pH 7 buffer (yellow)
- Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and colour-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
: Each lot compared to NIST Standard Reference Buffer.
Specifications
Specifications
- Pack type:Cubetainer®
- pH buffer type:Phthalate
- Pk:4 L
- Reference temperature:25 °C
- Colour:Red
- pH accuracy:±0,04
- pH value:4
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



