Order Entry
India
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

7, ±0,04, Phosphate, Yellow, 25 °C, Cubetainer®, 4 L
Catalog # 5656-03
Supplier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
Catalog # 5656-03
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded

7, ±0,04, Phosphate, Yellow, 25 °C, Cubetainer®, 4 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 4 L
Product Details & Documents

The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).

  • Colour coded
  • Potassium hydrogen phthalate based pH 4 buffer (red)
  • Phosphate based pH 7 buffer (yellow)
  • Borate based pH 10 buffer (blue)

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.

Solutions are available in both clear and colour-coded varieties.

A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

 : Each lot compared to NIST Standard Reference Buffer.

Specifications

Specifications

  • Pack type:
    Cubetainer®
  • pH buffer type:
    Phosphate
  • Pk:
    4 L
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • pH accuracy:
    ±0,04
  • pH value:
    7

Specifications