VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld
Product Details & Documents
These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter and no need to adjust the meter’s settings). The measured data is complete with serial number, settings and calibration data.
- One universal measuring channel
- Digital sensor recognition: The measured parameter and settings are recognised automatically
- Suitable for Intelligent Digital Probes (IDP)
- Suitable for measurements with cable length up to 100 metres
- 4500-point memory
- Easy to read backlit B/W graphic display
- Low energy consumption (1000 operating hours)
: Additional probes for other parameters can be added. Please contact your local Avantor sales office for further information on other electrodes available.
: 665-0494 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal IDP 7211 and USB cable.
665-0495 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml conductivity standard 0,01 mol/L KCl, conductivity cell IDP 761-C and USB cable.
665-0496 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, optical DO sensor OPOX11-3 and USB cable.
Specifications
- Description:Digital handheld phenomenal® MD 8000 H multi-parameter meter, pH kit, with intelligent gel plastic pH/temperature electrode IDP 711, 2 bottles of 50 ml pH buffers (1× pH 4,00 and 1× pH 7,00)
- Connections:1 measurement input
- Memory:4500 points
- Conductivity cell constant:Automatic
- IP rating:IP 67
- Data logging capacity:Up to 4500 data sets in automatic
- mV resolution:0,1; 1
- Resistivity range:1 Ωcm - 20 MΩcm (depending on sensor)
- Conductivity accuracy:±0,5% of value
- Temperature range:Auto: −5...+105 °C
- Salinity range:0 - 70
- mV range:±2500; ±1200
- Parameters:pH, mV, temperature, conductivity, dissolved oxygen, turbidity
- Reference temperature:20/25 °C
- pH range:0 - 14
- Calibration:Anycal 5-point customer buffer calibration
- Temperature resolution:0,1 °C
- DO concentration:0,00 - 20,00 / 0,0 - 50,0 mg/L
- Temperature accuracy:±0,2 K
- Interfaces:Mini USB-B (USB 1.1) for data downloads
- Cat. No.:665-0494
- pH accuracy:±0,1, ±0,01, ±0,005
- pH resolution:0,1/0,01/0,001
- Data logging - log function:Manual/time controlled
- Conductivity range:0,01 µS/cm - 1000 mS/cm (depending on sensor)
- TDS range:0 mg/L - 199,9 g/L
- mV accuracy:±0,3; ±1
- Calibration points:1 to 5 (automatic buffer recognition)
- DO saturation:0,0 - 200,0; 0 - 500 %
- Display:Backlit B/W graphic LCD
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



