Order Entry
India
ContactUsLinkComponent
VWR Single Channel Variable Volume Micropipette. Fully Autoclavable, Easy Volume Adjustment, Universal Tip Cone, Large 4 digit display, Large Grippy and Built-in Streamlined Tip Ejector. 1 * 1 items
Catalog # 613-6061
Supplier:  VWR Collection
VWR Single Channel Variable Volume Micropipette. Fully Autoclavable, Easy Volume Adjustment, Universal Tip Cone, Large 4 digit display, Large Grippy and Built-in Streamlined Tip Ejector. 1 * 1 items
Catalog # 613-6061
Supplier:  VWR Collection
Frequently Bought Together