Order Entry
India
ContactUsLinkComponent
Digital set-up for RPM + time with independent LED display, Maximum stirring speed upto 80RPM, Brushless DC motor, Compatible for us e with wide range of tubes, Used for prevention of blood coagulation and immune precipitation etc. Efficient Rolling 1 * 1 items
Catalog # 734-3247
Supplier:  VWR Collection
Digital set-up for RPM + time with independent LED display, Maximum stirring speed upto 80RPM, Brushless DC motor, Compatible for us e with wide range of tubes, Used for prevention of blood coagulation and immune precipitation etc. Efficient Rolling 1 * 1 items
Catalog # 734-3247
Supplier:  VWR Collection
Frequently Bought Together